Thermo Scientific Chemicals

Copper(II) chloride, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 035673.14
25 g, Each
Thermo Scientific Chemicals

Copper(II) chloride, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 035673.14
25 g, Each
Quantity
Catalog number: 035673.14
also known as 035673-14
Price (USD)
Price: 233.00
Online price: 198.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7447-39-4
IUPAC Name
copper(2+) dichloride
Molecular Formula
Cl2Cu
InChI Key
ORTQZVOHEJQUHG-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Cu++]
Appearance (Color)
Brown
Form
Powder
Residual water
< 100 ppm
Total Metal Impurities
0.005% max.

Description

Copper(II) chloride is used as a co-catalyst with palladium(II) chloride used in the Wacker process to prepare ethanol from ethane. It acts as an effective catalyst for the tetrahydropyranylation of alcohols. It is involved in the production of vinyl chloride and dichloroethane as well as in the chlorination of aromatic hydrocarbons. It acts as a deodorizing and desulfurizing agent in petroleum industry, mordant in textiles industry and in electrotyping baths for plating copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text