Antimony(III) iodide, ultra dry, 99.998% (metals basis)
Antimony(III) iodide, ultra dry, 99.998% (metals basis)
Antimony(III) iodide, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals

Antimony(III) iodide, ultra dry, 99.998% (metals basis)

CAS: 7790-44-5 | I3Sb | 502.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number 035676.03
also known as 035676-03
Price (USD)
50.65
Online Exclusive
56.40
Save 5.75 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.40
Save 5.75 (10%)
Each
Chemical Identifiers
CAS7790-44-5
IUPAC Nameantimony(3+) triiodide
Molecular FormulaI3Sb
InChI KeyKWQLUUQBTAXYCB-UHFFFAOYSA-K
SMILES[Sb+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water< 100 ppm
Total Metal Impurities0.002% max.
Particle Size-10 Mesh
Appearance (Color)Red
FormBeads
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only