Bismuth(III) chloride, ultra dry, 99.997+% (metals basis)
Bismuth(III) chloride, ultra dry, 99.997+% (metals basis)
Bismuth(III) chloride, ultra dry, 99.997+% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) chloride, ultra dry, 99.997+% (metals basis)

CAS: 7787-60-2 | BiCl3 | 315.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 035680.06
also known as 035680-06
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
55.50
Save 7.85 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
55.50
Save 7.85 (14%)
Each
Chemical Identifiers
CAS7787-60-2
IUPAC Namebismuth(3+) trichloride
Molecular FormulaBiCl3
InChI KeyJHXKRIRFYBPWGE-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.003% max.
FormPowder
Appearance (Color)White
Residual water< 100 ppm
Bismuth chloride is used as a catalyst in organic synthesis. In particular, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. The addition of other metal iodides increases its catalytic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only