Bismuth(III) iodide, ultra dry, 99.998% (metals basis)
Bismuth(III) iodide, ultra dry, 99.998% (metals basis)
Bismuth(III) iodide, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) iodide, ultra dry, 99.998% (metals basis)

CAS: 7787-64-6 | BiI3 | 589.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 035681.06
also known as 035681-06
Price (USD)
157.65
Online exclusive
175.00
Save 17.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
157.65
Online exclusive
175.00
Save 17.35
Each
Chemical Identifiers
CAS7787-64-6
IUPAC Namebismuth(3+) triiodide
Molecular FormulaBiI3
InChI KeyKOECRLKKXSXCPB-UHFFFAOYSA-K
SMILES[I-].[I-].[I-].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
Total Metal Impurities0.002% max.
FormGranules
Residual water< 100 ppm
Appearance (Color)Black
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only