Gallium(II) chloride, ultra dry, 99.999% (metals basis)
Gallium(II) chloride, ultra dry, 99.999% (metals basis)
Gallium(II) chloride, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Gallium(II) chloride, ultra dry, 99.999% (metals basis)

CAS: 24597-12-4
Have Questions?
Catalog NumberQuantityPackaging
035696.03
also known as 035696-03
1 gAmpule
Catalog number 035696.03
also known as 035696-03
Price (USD)
75.40
Each
Quantity:
1 g
Packaging:
Ampule
Request bulk or custom format
Price (USD)
75.40
Each
Chemical Identifiers
CAS24597-12-4
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Residual water< 100 ppm
Appearance (Color)White
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only