Gallium(III) iodide, ultra dry, 99.999% (metals basis)
Gallium(III) iodide, ultra dry, 99.999% (metals basis)
Gallium(III) iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Gallium(III) iodide, ultra dry, 99.999% (metals basis)

CAS: 13450-91-4 | GaI3 | 450.436 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 035699.03
also known as 035699-03
Price (USD)
63.65
Online Exclusive
71.00
Save 7.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
71.00
Save 7.35 (10%)
Each
Chemical Identifiers
CAS13450-91-4
IUPAC Namegallium(3+) triiodide
Molecular FormulaGaI3
InChI KeyDWRNSCDYNYYYHT-UHFFFAOYSA-K
SMILES[Ga+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities0.001% max.
Residual water< 100 ppm
Appearance (Color)Yellow
Gallium(III) iodide is widely used as lewis acid, a precursor of organogallium reagents. It can also be involved in detection of solar neutrinos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only