Indium(I) iodide, ultra dry, 99.998% (metals basis)
Indium(I) iodide, ultra dry, 99.998% (metals basis)
Indium(I) iodide, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals

Indium(I) iodide, ultra dry, 99.998% (metals basis)

CAS: 13966-94-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 035700.03
also known as 035700-03
Price (USD)
90.65
Online exclusive
100.00
Save 9.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
90.65
Online exclusive
100.00
Save 9.35
Each
Chemical Identifiers
CAS13966-94-4
IUPAC Nameλ¹-indiganyliumylidene iodide
Molecular FormulaIIn
InChI KeyFOVZCYAIUZHXGB-UHFFFAOYSA-M
SMILES[In+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
Appearance (Color)Red to brown
Residual water< 100 ppm
Total Metal Impurities0.002% max.
FormBeads
Indium(I) iodide can be used to prepare indium cluster and chain compounds. It is also used as a reagent to propagate organic reactions viz. cyclization and cleavage of diselenide or disulfide. It plays an important role in preparing indium containing complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only