Magnesium chloride, ultra dry, 99.9% (metals basis)
Magnesium chloride, ultra dry, 99.9% (metals basis)
Magnesium chloride, ultra dry, 99.9% (metals basis)
Thermo Scientific Chemicals

Magnesium chloride, ultra dry, 99.9% (metals basis)

Widely used as a source of magnesium ion, co-factor for many enzymes, for dust control and road stabilization. Also used in wastewater treatment, wallboard, artificial seawater, feed supplement, textiles, paper, fireproofing agents, cements and refrigeration brine. | CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 035707.09
also known as 035707-09
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.70
Save 12.05 (14%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.70
Save 12.05 (14%)
Each
Chemical Identifiers
CAS7786-30-3
IUPAC Namemagnesium(2+) dichloride
Molecular FormulaCl2Mg
InChI KeyTWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBead
Total Metal Impurities0.1% max.
Particle Size-10 Mesh
It is widely used as a source of magnesium ion, co-factor for many enzymes, for dust control and road stabilization. Also used in wastewater treatment, wallboard, artificial seawater, feed supplement, textiles, paper, fireproofing agents, cements and refrigeration brine. It is an important coagulant, an elution buffer for antibody affinity column purifications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only