Nickel(II) chloride, ultra dry, 99.9% (metals basis)
Nickel(II) chloride, ultra dry, 99.9% (metals basis)
Nickel(II) chloride, ultra dry, 99.9% (metals basis)
Thermo Scientific Chemicals

Nickel(II) chloride, ultra dry, 99.9% (metals basis)

CAS: 7718-54-9 | Cl2Ni | 129.593 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 035713.06
also known as 035713-06
Price (USD)
131.65
Online exclusive
146.00
Save 14.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
131.65
Online exclusive
146.00
Save 14.35
Each
Chemical Identifiers
CAS7718-54-9
IUPAC Namenickel(2+) dichloride
Molecular FormulaCl2Ni
InChI KeyQMMRZOWCJAIUJA-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Residual water< 100 ppm
Total Metal Impurities0.1% max.
FormPowder
It is used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only