Cobalt(II) fluoride, ultra dry, 99.99% (metals basis)
Cobalt(II) fluoride, ultra dry, 99.99% (metals basis)
Cobalt(II) fluoride, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Cobalt(II) fluoride, ultra dry, 99.99% (metals basis)

CAS: 10026-17-2 | CoF2 | 96.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 035740.09
also known as 035740-09
Price (USD)
216.65
Online Exclusive
241.00
Save 24.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
216.65
Online Exclusive
241.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS10026-17-2
IUPAC Nameλ²-cobalt(2+) difluoride
Molecular FormulaCoF2
InChI KeyYCYBZKSMUPTWEE-UHFFFAOYSA-L
SMILES[F-].[F-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water< 100 ppm
Appearance (Color)Pink to red
Total Metal Impurities0.01% max.
FormPowder
It is used as a catalyst to alloy metals and also used for optical deposition, of which it tremendously improves optical quality, in dental care.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only