Chromium(III) fluoride, 98%
Chromium(III) fluoride, 98%
Chromium(III) fluoride, 98%
Chromium(III) fluoride, 98%
Thermo Scientific Chemicals

Chromium(III) fluoride, 98%

CAS: 7788-97-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 035741.04
also known as 035741-04
Price (USD)
157.65
Online Exclusive
175.00
Save 17.35 (10%)
Each
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)
157.65
Online Exclusive
175.00
Save 17.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7788-97-8
IUPAC Namechromium trifluoride
Molecular FormulaCrF3
InChI KeyQZEZRQVFRMLNPK-UHFFFAOYSA-N
SMILES[F].[F].[F].[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA% Fluoride: ≥51.0% (equiv. to ≥97.5% purity)
Appearance (Color)Powder or crystals or crystalline powder
Appearance (Color)Green
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only