Yttrium(III) iodide, ultra dry, 99.9% (REO)
Yttrium(III) iodide, ultra dry, 99.9% (REO)
Yttrium(III) iodide, ultra dry, 99.9% (REO)
Thermo Scientific Chemicals

Yttrium(III) iodide, ultra dry, 99.9% (REO)

CAS: 13470-38-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 035744.03
also known as 035744-03
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS13470-38-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormPowder/chunks
Total Rare Earth Oxide Impurities≤0.1% (≤1000ppm)
Yttrium(III) iodide, is used as a starting material for YBCO superconducting materials in a lower temperature preparation. Applied in the solid-state synthesis of a new compound, Y21I18C14B7, which is the first example of a rare earth metal halide with interstitial halogen atoms with misfit characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only