Yttrium(III) bromide, ultra dry, 99.9% (REO)
Yttrium(III) bromide, ultra dry, 99.9% (REO)
Yttrium(III) bromide, ultra dry, 99.9% (REO)
Thermo Scientific Chemicals

Yttrium(III) bromide, ultra dry, 99.9% (REO)

CAS: 13469-98-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 035746.04
also known as 035746-04
Price (USD)
135.65
Online Exclusive
150.00
Save 14.35 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
135.65
Online Exclusive
150.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS13469-98-2
IUPAC Nameyttrium(3+) tribromide
Molecular FormulaBr3Y
InChI KeyFSDCGXUNLWDJNL-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.1% max.

Yttrium(III) bromide is used in electroplating, as catalyst, material enhancing & medical treatments in curing colds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only