Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL
Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL
Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL
Thermo Scientific Chemicals

Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL

CAS: 7439-88-5 | Ir | 192.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 035752.AD
also known as 035752-AD
Price (USD)
-
Quantity:
50 mL
Request bulk or custom format
Specifications
CAS7439-88-5
ChEBICHEBI:49666
Chemical Name or MaterialIridium
DOT InformationHazard Class: 8, Packing Group: II
EINECS Number231-595-7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only