Tungsten, AAS standard solution, Specpure™ W 1000μg/mL
Tungsten, AAS standard solution, Specpure™ W 1000μg/mL
Tungsten, AAS standard solution, Specpure™ W 1000μg/mL
Thermo Scientific Chemicals

Tungsten, AAS standard solution, Specpure™ W 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 035760.AP
also known as 035760-AP
Price (USD)
70.20
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
70.20
Each
Specifications
Chemical Name or MaterialTungsten AAS standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H302-H312-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only