Ruthenium, plasma standard solution, Specpure™ Ru 1000μg/mL
Ruthenium, plasma standard solution, Specpure™ Ru 1000μg/mL
Ruthenium, plasma standard solution, Specpure™ Ru 1000μg/mL
Thermo Scientific Chemicals

Ruthenium, plasma standard solution, Specpure™ Ru 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 035767.AE
also known as 035767-AE
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
161.00
Save 24.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
161.00
Save 24.35 (15%)
Each
Specifications
Chemical Name or MaterialRuthenium plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID
EINECS Number231-595-7
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Ruthenium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only