Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL
Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL
Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL
Thermo Scientific Chemicals

Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL

W in 5% HNO3/tr. HF
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 035772.AP
also known as 035772-AP
Price (USD)
259.65
Online Exclusive
288.00
Save 28.35 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
259.65
Online Exclusive
288.00
Save 28.35 (10%)
Each
Specifications
Chemical Name or MaterialTungsten, plasma standard solution, Specpure™
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H302-H312-H332;
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only