Strontium carbonate, 99.99% (metals basis)
Strontium carbonate, 99.99% (metals basis)
Strontium carbonate, 99.99% (metals basis)
Strontium carbonate, 99.99% (metals basis)
Thermo Scientific Chemicals

Strontium carbonate, 99.99% (metals basis)

CAS: 1633-05-2 | CO3Sr | 147.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 035793.22
also known as 035793-22
Price (USD)
311.65
Special offer
Online exclusive
Ends: 28-Dec-2026
367.00
Save 55.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
311.65
Special offer
Online exclusive
Ends: 28-Dec-2026
367.00
Save 55.35
Each
Chemical Identifiers
CAS1633-05-2
IUPAC Namestrontium(2+) carbonate
Molecular FormulaCO3Sr
InChI KeyLEDMRZGFZIAGGB-UHFFFAOYSA-L
SMILES[Sr++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Used as a colorant in fireworks. It is also used in ceramics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only