Lanthanum(III) bromide, ultra dry, 99.9% (REO)
Lanthanum(III) bromide, ultra dry, 99.9% (REO)
Lanthanum(III) bromide, ultra dry, 99.9% (REO)
Thermo Scientific Chemicals

Lanthanum(III) bromide, ultra dry, 99.9% (REO)

CAS: 13536-79-3 | Br3La | 378.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 035800.18
also known as 035800-18
Price (USD)
1,128.65
Online Exclusive
1,254.00
Save 125.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
1,128.65
Online Exclusive
1,254.00
Save 125.35 (10%)
Each
Chemical Identifiers
CAS13536-79-3
IUPAC Namelanthanum(3+) tribromide
Molecular FormulaBr3La
InChI KeyXKUYOJZZLGFZTC-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.1% max.
Particle Size-10 Mesh
FormBeads

It is used as a scintillation material in certain application, as a source in the synthesis of lanthanum & its derivatives CsNaLa6Br14N2 and La3Br6N which contain rare bitetrahedral structural units..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only