Nickel Thinfoil, 0.0125mm (0.000492in) thick, 99% (metals basis excluding Co), Co nominal 0.8%
Nickel Thinfoil, 0.0125mm (0.000492in) thick, 99% (metals basis excluding Co), Co nominal 0.8%
Nickel Thinfoil, 0.0125mm (0.000492in) thick, 99% (metals basis excluding Co), Co nominal 0.8%
Thermo Scientific Chemicals

Nickel Thinfoil, 0.0125mm (0.000492in) thick, 99% (metals basis excluding Co), Co nominal 0.8%

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
Catalog number 035814.GH
also known as 035814-GH
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis excluding Co), Co nominal 0.8%
CAS7440-02-0
ChEBICHEBI:28112
Chemical Name or MaterialNickel Thinfoil
EINECS Number231-111-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only