Nickel Thinfoil, 0.01mm (0.0004in) thick, 99.95% (metals basis)
Nickel Thinfoil, 0.01mm (0.0004in) thick, 99.95% (metals basis)
Nickel Thinfoil, 0.01mm (0.0004in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Nickel Thinfoil, 0.01mm (0.0004in) thick, 99.95% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 035817.FF
also known as 035817-FF
Price (USD)
55.65
Online Exclusive
61.40
Save 5.75 (9%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.40
Save 5.75 (9%)
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
FormFoil, 0.01mm (0.0004in) thick.
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only