Cerium silicide, 99.9% (REO)
Cerium silicide, 99.9% (REO)
Cerium silicide, 99.9% (REO)
Cerium silicide, 99.9% (REO)
Cerium silicide, 99.9% (REO)
Cerium silicide, 99.9% (REO)
Cerium silicide, 99.9% (REO)
Cerium silicide, 99.9% (REO)
Cerium silicide, 99.9% (REO)
Thermo Scientific Chemicals

Cerium silicide, 99.9% (REO)

CAS: 12014-85-6 | CeSi2 | 196.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 035836.06
also known as 035836-06
Price (USD)
149.65
Special offer
Online exclusive
Ends: 30-Jun-2026
176.00
Save 26.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
149.65
Special offer
Online exclusive
Ends: 30-Jun-2026
176.00
Save 26.35 (15%)
Each
Chemical Identifiers
CAS12014-85-6
IUPAC Nameλ⁴-cerium(4+) disil-1-ene-1,1,2,2-tetraide
Molecular FormulaCeSi2
InChI KeyKLTZYOJHFGPJMO-UHFFFAOYSA-N
SMILES[Ce+4].[Si--]=[Si--]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPieces, 3-15mm
Appearance (Color)Grey
Total Rare Earth Oxide Impurities≤0.1%
Cerium silicide is used as a surrogate material to investigate the feasibility of high-energy ball milling and Spark Plasma Sintering (SPS) for the production of uranium silicide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only