Bis(cyclopentadienyl)hafnium dichloride
Bis(cyclopentadienyl)hafnium dichloride
Bis(cyclopentadienyl)hafnium dichloride
Thermo Scientific Chemicals

Bis(cyclopentadienyl)hafnium dichloride

CAS: 12116-66-4 | C10H10Cl2Hf | 379.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036105.14
also known as 036105-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Specifications
CAS12116-66-4
Chemical Name or MaterialBis(cyclopentadienyl)hafnium dichloride
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECS Number235-177-5
Formula Weight379.59
View more
Starting material to prepare many kinds of hafnocene(II or IV) derivatives; in combination with AgI salt, is an effective activator for glycosyl fluoride. Pharmaceutical intermediates and alos used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only