Sodium selenide, 99.8% (metals basis)
Sodium selenide, 99.8% (metals basis)
Sodium selenide, 99.8% (metals basis)
Sodium selenide, 99.8% (metals basis)
Thermo Scientific Chemicals

Sodium selenide, 99.8% (metals basis)

CAS: 1313-85-5 | Na2Se | 124.951 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 036187.03
Price (USD)
361.65
Special offer
Online exclusive
Ends: 30-Jun-2026
425.00
Save 63.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
361.65
Special offer
Online exclusive
Ends: 30-Jun-2026
425.00
Save 63.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1313-85-5
IUPAC Namedisodium selandiide
Molecular FormulaNa2Se
InChI KeyVPQBLCVGUWPDHV-UHFFFAOYSA-N
SMILES[Na+].[Na+].[Se--]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Sodium selenide is used in paper and pulp industry, for kraft process followed by photographic industry to prevent developer solutions. It finds application as a bleaching agent in textile industry, sulfur dyes and detergents. It is also used to impart color to glass, glazes and enamels. Further, it acts as a decolorizer of glass due to neutralization of greenish tint produced by iron (ferrous) compounds. It plays an important role to prepare organoselenium compounds by reacting with alkyl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only