Ammonium vanadium oxide, ACS, 99.0% min
Ammonium vanadium oxide, ACS, 99.0% min
Ammonium vanadium oxide, ACS, 99.0% min
Thermo Scientific Chemicals

Ammonium vanadium oxide, ACS, 99.0% min

CAS: 7803-55-6 | H4NO3V | 116.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 036213.18
also known as 036213-18
Price (USD)
55.50
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
55.50
Each
Chemical Identifiers
CAS7803-55-6
IUPAC Namevanadiumoylol amine
Molecular FormulaH4NO3V
InChI KeyYBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILESN.O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% NH₄VO₃ min
Impurity contentCO₃ P.T. (about 0.3%)
Impurity contentSO₄ 0.05%
Solution Testin ammonium hydroxide P.T.
Impurity contentCl 0.2%
Ammonium vanadium oxide is used as a catalyst in organic synthesis, and as an analytical reagent in laboratories. It is employed in the preparation of vanadate. It acts as a sensor for the determination of N-acetyltransferase activity in humans. Further it finds use in dyes, varnishes, indelible inks, drier for paints, and in photography. With copper carbonate, it has been shown to enhance the catalytic activities. Ammonium metavanadate is an environmental-friendly catalyst to prepare alpha-hydroxyphosphonate derivatives in high yield through the reaction of aryl/heteroaryl aldehydes with triethylphosphonate derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only