Search
View additional product information for Lithium chloride, ACS, 99% min - FAQs (036217.36, 036217.A3)
1 product FAQs found
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.