Chromium(III) oxide, 99.6% (metals basis)
Chromium(III) oxide, 99.6% (metals basis)
Chromium(III) oxide, 99.6% (metals basis)
Thermo Scientific Chemicals

Chromium(III) oxide, 99.6% (metals basis)

CAS: 1308-38-9 | Cr2O3 | 151.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 036258.22
also known as 036258-22
Price (USD)
228.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
228.00
Each
Chemical Identifiers
CAS1308-38-9
IUPAC Namedichromium(3+) trioxidandiide
Molecular FormulaCr2O3
InChI KeyUOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Cr+3].[Cr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.4% max.
For vacuum deposition. It can be used in selective oxidation. It is mainly used for smelting metal chromium and chromium carbide. It is used as coloring agent for color painting on enamelware and porcelain ware, imitation leather or building material. It is also used for making sun-proof coating, abrasive, green polishing compound and special printing ink for printing bank note. It is used as catalyst for organic synthetic chromium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only