Antimony(III) telluride, 99.999% (metals basis)
Antimony(III) telluride, 99.999% (metals basis)
Antimony(III) telluride, 99.999% (metals basis)
Thermo Scientific Chemicals

Antimony(III) telluride, 99.999% (metals basis)

For vacuum deposition. Activator in glass industry. Paints and Adhesives industries. Pigments, Ceramic frites. As semiconductor material, it has a large thermoelectric effect and is therefore used in solid state refrigerators. | CAS: 1327-50-0 | Sb2Te3 | 626.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 036282.09
also known as 036282-09
Price (USD)
161.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
161.00
Each
Chemical Identifiers
CAS1327-50-0
IUPAC Namebis(tellanylidenestibanyl) tellurium
Molecular FormulaSb2Te3
InChI KeyBPDQXJZWVBPDSN-UHFFFAOYSA-N
SMILES[Te].[Sb]=[Te].[Sb]=[Te]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
For vacuum deposition. Activator in glass industry. Paints and Adhesives industries. Pigments, Ceramic frites. As semiconductor material, it has a large thermoelectric effect and is therefore used in solid state refrigerators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only