4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%
Thermo Scientific Chemicals

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%

CAS: 116-63-2 | C10H9NO4S | 239.245 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 036290.06
also known as 036290-06
Price (USD)
27.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
27.00
Each
Chemical Identifiers
CAS116-63-2
IUPAC Name4-amino-3-hydroxynaphthalene-1-sulfonic acid
Molecular FormulaC10H9NO4S
InChI KeyRXCMFQDTWCCLBL-UHFFFAOYSA-N
SMILESNC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solution Testin sodium carbonate P.T.
CommentSensitivity to phosphate P.T.
Impurity contentSO₄ 0.2%
Residue on ignition0.1%
4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as a key starting material for the preparation of naphthalenesulfonic acid derivatives by reacting with palmitoyl chloride, which shows the activity to inhibit DNA polymerase and ribonuclease H Activities of HIV-1 reverse transcriptase. It is also used to prepare a copolyner with melamine, which is associated with graphene to prepare molecularly imprinted sensor and utilized for the determination of melatonin in human biological fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only