Aluminum shot, ACS, 8-12mm (0.31-0.47in)
Aluminum shot, ACS, 8-12mm (0.31-0.47in)
Aluminum shot, ACS, 8-12mm (0.31-0.47in)
Thermo Scientific Chemicals

Aluminum shot, ACS, 8-12mm (0.31-0.47in)

Typically 3-4mm thick | CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 036293.22
also known as 036293-22
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.20
Save 9.55 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.20
Save 9.55 (15%)
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentMn 0.002%
Impurity contentTi 0.03%
Impurity contentCu 0.02%
Impurity contentFe 0.1%
Impurity contentInsoluble in dilute hydrochloric acid 0.05%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only