Nickel Chromium lump, 99.9% (metals basis)
Nickel Chromium lump, 99.9% (metals basis)
Nickel Chromium lump, 99.9% (metals basis)
Thermo Scientific Chemicals

Nickel Chromium lump, 99.9% (metals basis)

For vacuum deposition | CAS: 11106-97-1 | CrNi | 110.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 036298.14
also known as 036298-14
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS11106-97-1
IUPAC Namechromium nickel
Molecular FormulaCrNi
InChI KeyVNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES[Cr].[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormLump, 3-12mm
Assay from Supplier's CofA≥99.9% (metals basis)
Assay from Supplier's CofANi:Cr; 80:20 wt%
For vacuum deposition. hey are also used in some dental restorations. Used as Heating Elements, Thermocouples, Wear Resistant Alloys. Used as intermediate and research. Nichrome wire is commonly used in ceramics and Pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only