Azelaic acid, 98%
Azelaic acid, 98%
Azelaic acid, 98%
Thermo Scientific Chemicals

Azelaic acid, 98%

CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
5 x 100 g
Catalog number 036308.14
also known as 036308-14
Price (USD)
38.65
Online exclusive
42.90
Save 4.25
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
38.65
Online exclusive
42.90
Save 4.25
Each
Chemical Identifiers
CAS123-99-9
IUPAC Namenonanedioic acid
Molecular FormulaC9H16O4
InChI KeyBDJRBEYXGGNYIS-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Assay (Titration)>97.5%
FormPowder or flakes
Useful in lubrication and plasticizers. Azelaic acid is used to treat acne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only