Nickel(II) sulfate hexahydrate, ACS, 98.0% min
Nickel(II) sulfate hexahydrate, ACS, 98.0% min
Nickel(II) sulfate hexahydrate, ACS, 98.0% min
Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, ACS, 98.0% min

CAS: 10101-97-0 | H12NiO10S | 262.839 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 036336.A4
Price (USD)
348.65
Special offer
Online exclusive
Ends: 30-Jun-2026
410.00
Save 61.35 (15%)
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
348.65
Special offer
Online exclusive
Ends: 30-Jun-2026
410.00
Save 61.35 (15%)
Each
Chemical Identifiers
CAS10101-97-0
IUPAC Namenickel(2+) hexahydrate sulfate
Molecular FormulaH12NiO10S
InChI KeyRRIWRJBSCGCBID-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCo 0.002%
Impurity contentCu 0.005%
Impurity contentFe 0.001%
Impurity contentK 0.01%
Impurity contentMg 0.005%
View more
It is the precursor material for Ni electroplating. It is applied in biochemistry and also employed as a calibrant for magnetic susceptibility measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only