Copper wire, 0.5mm (0.02in) dia, Puratronic™, 99.999% (metals basis), Oxygen free
Copper wire, 0.5mm (0.02in) dia, Puratronic™, 99.999% (metals basis), Oxygen free
Copper wire, 0.5mm (0.02in) dia, Puratronic™, 99.999% (metals basis), Oxygen free
Thermo Scientific Chemicals

Copper wire, 0.5mm (0.02in) dia, Puratronic™, 99.999% (metals basis), Oxygen free

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
250 cm
Catalog number 036397.BW
also known as 036397-BW
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.90
Save 13.25 (15%)
Each
Quantity:
50 cm
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 30-Jun-2026
87.90
Save 13.25 (15%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormWire, 0.5mm (0.02in) diameter.
Assay≥99.999% (metals basis)
Impurity contentOxygen: ≤5 ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only