N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
Thermo Scientific Chemicals

N,N-Dimethyl-1-naphthylamine, 99%

CAS: 86-56-6 | C12H13N | 171.243 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number 036438.AA
also known as 036438-AA
Price (USD)
38.00
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
38.00
Each
Specifications
CAS86-56-6
Chemical Name or MaterialN,N-Dimethyl-1-naphthylamine
ColorYellow
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Density1.039 g/mL
View more
N,N-Dimethyl-1-naphthylamine is used in the nitrate reductase test to form a red precipitate of Prontosil by reacting with a nitrite-sulfanilic acid complex. And is also used to detection and determination of nitrites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only