Sodium bromide, ACS, 99.0% min
Sodium bromide, ACS, 99.0% min
Sodium bromide, ACS, 99.0% min
Thermo Scientific Chemicals

Sodium bromide, ACS, 99.0% min

CAS: 7647-15-6 | BrNa | 102.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 036444.A3
also known as 036444-A3
Price (USD)
252.00
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
252.00
Each
Chemical Identifiers
CAS7647-15-6
IUPAC Namesodium bromide
Molecular FormulaBrNa
InChI KeyJHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES[Na+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% NaBr min
Impurity contentBrO₃ 0.001%
Impurity contentCl 0.2%
Impurity contentBa 0.002%
Impurity contentCa 0.002%
View more
It is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only