Hexamethylenetetramine, ACS, 99+%
Hexamethylenetetramine, ACS, 99+%
Hexamethylenetetramine, ACS, 99+%
Thermo Scientific Chemicals

Hexamethylenetetramine, ACS, 99+%

CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036462.22
also known as 036462-22
Price (USD)
32.70
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
32.70
Each
Chemical Identifiers
CAS100-97-0
IUPAC Name1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Molecular FormulaC6H12N4
InChI KeyVKYKSIONXSXAKP-UHFFFAOYSA-N
SMILESC1N2CN3CN1CN(C2)C3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Residue on ignition≤0.1%
Loss on Drying≤2.0%
Heavy Metals (as Pb)≤0.001%
Assay (Titration)≥99.0% (dried basis)
View more
Hexamethylenetetramine is used in the manufacturing of phenolic resins liquid preparations and molding compounds. It finds applications in in brake and clutch linings, abrasive products, non-woven textiles and fireproof materials. It is also used as a drug taken by mouth to prevent recurrent urinary tract infections. It is used as a solid fuel in association with 1,3,5-trioxane in campers, hobbyists, the military and relief organizations. It acts as a food additive and as a preservative. It is an important reagent in organic synthesis, especially Duff reaction, Sommelet reaction and in the Delepine reaction. Further, it is used in adhesives, coatings, sealants, and leather preservatives. It is utilized as a formaldehyde releaser for photographic film development and as a hardener for epoxy resins of the bisphenol A type.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only