Ethanol-d{6}, 99.5%(Isotopic)
Ethanol-d{6}, 99.5%(Isotopic)
Ethanol-d{6}, 99.5%(Isotopic)
Thermo Scientific Chemicals

Ethanol-d{6}, 99.5%(Isotopic)

CAS: 1516-08-1 | C2H6O | 52.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036502.03
also known as 036502-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1516-08-1
IUPAC Name(²H₅)ethan-1-(²H)ol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-LIDOUZCJSA-N
SMILES[2H]OC([2H])([2H])C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.5% Isotopic enrichment by ¹H nmr
ClarityClear
Water Content (Karl Fischer Titration)≤1000ppm
Appearance (Color)Colorless
Assay (GC)≥99%
View more
Persimmon, the fruit treated with ethanol-d_6, thioethers 9d and 9e were obtained, indicating that the C_2 unit originated from acetaldehyde which generated by oxidation of ethanol added externally.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only