Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
Thermo Scientific Chemicals

Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 036526.36
also known as 036526-36
Price (USD)
93.50
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
93.50
Each
Chemical Identifiers
CAS13446-34-9
IUPAC Namemanganese(2+) tetrahydrate dichloride
Molecular FormulaCl2H8MnO4
InChI KeyCNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILESO.O.O.O.[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentSO₄ 0.005%
Assay (unspecified)98.0-101.0% Cl₂Mn.4H₂O
Impurity contentCa 0.005%
Impurity contentFe 5ppm
Impurity contentK 0.01%
View more
It is used in the production of dry cell batteries. It is a precursor material for antiknock compound methylcyclopentadienyl manganese tricarbony. It is also useful for coupling reactions as well as organic synthesis. It is a catalyst for fast and slow dehydrogenation of ball milled lithium alanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only