Pararosaniline hydrochloride, 90+%, Thermo Scientific Chemicals
Pararosaniline hydrochloride, 90+%, Thermo Scientific Chemicals
Pararosaniline hydrochloride, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pararosaniline hydrochloride, 90+%, Thermo Scientific Chemicals

pH indicator 1.2 (purple) to 3.0 (red)
Have Questions?
Catalog NumberQuantity
036540.18
also known as 036540-18
50 g
Catalog number 036540.18
also known as 036540-18
Price (USD)
139.65
Online Exclusive
155.00
Save 15.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
139.65
Online Exclusive
155.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS569-61-9
IUPAC Namehydrogen 4-[(4-aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular FormulaC19H18ClN3
InChI KeyJUQPZRLQQYSMEQ-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)=C1C=CC(=N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
UV/VisUV Ratios: Report
Assay (Dye)≥90%
Appearance (Color)Dark green
Pararosaniline hydrochloride is used to dye polyacrylonitrile fibers. It is also is used as a colorimetric test for aldehydes, in the Schiff test. It is the only basic fuchsine component suitable for making the aldehyde-fuchsine stain for pancreatic islet beta cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only