Tetraethylammonium hydroxide, 35% w/w aq. soln.
Tetraethylammonium hydroxide, 35% w/w aq. soln.
Tetraethylammonium hydroxide, 35% w/w aq. soln.
Thermo Scientific Chemicals

Tetraethylammonium hydroxide, 35% w/w aq. soln.

CAS: 77-98-5 | C8H21NO | 147.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 036560.22
also known as 036560-22
Price (USD)
56.65
Special offer
Online exclusive
Ends: 28-Dec-2026
66.70
Save 10.05
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 28-Dec-2026
66.70
Save 10.05
Each
Chemical Identifiers
CAS77-98-5
IUPAC Nametetraethylazanium hydroxide
Molecular FormulaC8H21NO
InChI KeyLRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES[OH-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤100
Assay (Aqueous acid-base Titration)≥32.5 to ≤37.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Refractive Index1.4040-1.4070 @ 20°C (non-U.S. specification)
FormLiquid
View more
Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only