Niobium n-butoxide, 99% (metals basis)
Niobium n-butoxide, 99% (metals basis)
Niobium n-butoxide, 99% (metals basis)
Niobium n-butoxide, 99% (metals basis)
Thermo Scientific Chemicals
Niobium n-butoxide, 99% (metals basis)
CAS: 51030-47-8 | C20H50NbO5 | 463.521 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 036570.03
also known as 036570-03
Price (USD)
60.80
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
60.80
Each
Add to cart
Chemical Identifiers
CAS51030-47-8
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Niobium n-butoxide plays an important role as a precursor in chemical synthesis. It is also used in the production of nano-materials. It acts as a precursor for dielectric films and structures with controlled properties.