This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036573.AC
also known as 036573-AC
Price (USD)
126.00
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
126.00
Each
Specifications
Assay Percent Notes(REO)
CAS19236-15-8
Chemical Name or MaterialNeodymium(III) isopropoxide
ColorBlue
Concentration or Composition (by Analyte or Components)2.5% w/v in toluene/isopropanol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.