Zinc telluride, 99.998% (metals basis)
Zinc telluride, 99.998% (metals basis)
Zinc telluride, 99.998% (metals basis)
Thermo Scientific Chemicals

Zinc telluride, 99.998% (metals basis)

CAS: 1315-11-3 | TeZn | 192.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 036595.18
also known as 036595-18
Price (USD)
588.65
Special offer
Online exclusive
Ends: 30-Jun-2026
692.00
Save 103.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
588.65
Special offer
Online exclusive
Ends: 30-Jun-2026
692.00
Save 103.35 (15%)
Each
Chemical Identifiers
CAS1315-11-3
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Zinc telluride is a semiconductor material used in optoelectronics. It is used in blue LEDs, laser diodes, solar cells, components of microwave generators, electro-optic detector, holographic interferometry and laser optical phase conjugation devices. It is associated with lithium niobate and used for the generation of pulsed terahertz radiation in time-domain terahertz spectroscopy and terahertz imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only