Formic acid, ACS, 96+%
Formic acid, ACS, 96+%
Formic acid, ACS, 96+%
Formic acid, ACS, 96+%
Thermo Scientific Chemicals

Formic acid, ACS, 96+%

CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
4 x 500 g
Catalog number 036617.36
also known as 036617-36
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.20
Save 13.55 (15%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.20
Save 13.55 (15%)
Each
Add to cart
Chemical Identifiers
CAS64-18-6
IUPAC Nameformic acid
Molecular FormulaCH2O2
InChI KeyBDAGIHXWWSANSR-UHFFFAOYSA-N
SMILESOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride content0.001%, max
Heavy Metals0.001%, max ( as Pb)
Color (APHA)15, max
Impurity contentSO₄ 0.003%, max
CommentDilution Test P.T.
View more
Used as a hydrogen source, as a reducing agent and is used in tanning and dye fixing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only