Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis)
Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis)
Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis)
Thermo Scientific Chemicals

Yttrium(III) 2-ethylhexanoate, 99.8% (metals basis)

CAS: 103470-68-4 | C24H45O6Y | 518.524 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 036661.14
also known as 036661-14
Price (USD)
152.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
152.00
Each
Specifications
Assay Percent Notes(metals basis)
CAS103470-68-4
Chemical Name or MaterialYttrium(III) 2-ethylhexanoate
Formula Weight518.16
Health Hazard 1H361d
View more
Yttrium(III) 2-ethylhexanoate has been used for research in nanotechnology based applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only