Methyl Red hydrochloride, ACS
Methyl Red hydrochloride, ACS
Methyl Red hydrochloride, ACS
Thermo Scientific Chemicals

Methyl Red hydrochloride, ACS

CAS: 63451-28-5 | C15H16ClN3O2 | 305.762 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 036668.22
also known as 036668-22
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.60
Save 12.95 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.60
Save 12.95 (14%)
Each
Chemical Identifiers
CAS63451-28-5
IUPAC Namehydrogen 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid chloride
Molecular FormulaC15H16ClN3O2
InChI KeyICTWKXAVJAHRMK-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentClarity of alcohol solution P.T.
CommentVisual transition interval from pH 4.2 (pink) to 6.2 (yellow)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only