Thermo Scientific Chemicals

Methyl-p-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog number: 036670.09
10 g, Each
Thermo Scientific Chemicals

Methyl-p-benzoquinone, 98%, Thermo Scientific Chemicals

Catalog number: 036670.09
10 g, Each
Quantity
Catalog number: 036670.09
also known as 036670-09
Price (USD)
Price: 37.80
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
553-97-9
Appearance (Color)
Yellow to brown
Assay (GC)
≥97.5%
Form
Crystalline powder
Identification (FTIR)
Conforms
Proton NMR
Conforms

Description

Methyl-p-benzoquinone imine is the products were the tetrahedral adducts. Synthesis of 2H-indazole-4,7-dione derivatives from3-phenylsydnone and p-toluquinone. A Michael addition of 2-methylcyclopentane-1,3-dione to various p-toluquinone imides exclusively afforded the sole products in a regiospecific manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text