4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%
4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%
4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%
Thermo Scientific Chemicals

4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%

CAS: 55-55-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 036674.30
also known as 036674-30
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS55-55-0
IUPAC Namebis(4-(methylamino)phenol); sulfuric acid
Molecular FormulaC14H20N2O6S
InChI KeyZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSuitability for determination of phosphate P.T.
Assay (Titration)99.0-101.5%
Residue on ignition<0.1%
FormSolid (discolors in air)
4-(Methylamino)phenol sulfate is a photographic developer used in black and white photography. It is also used as tone developers in developing agents. Further, it is used in oxidative hair dye formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only