Potassium permanganate, low in mercury, ACS, 99.0% min
Potassium permanganate, low in mercury, ACS, 99.0% min
Potassium permanganate, low in mercury, ACS, 99.0% min
Thermo Scientific Chemicals

Potassium permanganate, low in mercury, ACS, 99.0% min

CAS: 7722-64-7 | KMnO4 | 158.032 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 036675.A4
also known as 036675-A4
Price (USD)
303.00
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
303.00
Each
Chemical Identifiers
CAS7722-64-7
IUPAC Namepotassium manganesoylolate
Molecular FormulaKMnO4
InChI KeyVZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES[K+].[O-][Mn](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% KMnO₄ min
Impurity contentHg 0.05ppm
Insoluble material0.2%
Impurity contentChloride and chlorate (as Cl) 0.005%
Impurity contentSO₄ 0.02%
Potassium permanganate is an oxidizing agent, is used in the synthesis of organic compounds like ascorbic acid, chloramphenicol, saccharin, isonicotinic acid and pyrazinoic acid and in pharmaceutical intermediate. It is used to remove ferrous impurities from inorganic chemicals. It is also used in analytical chemistry as a reagent for redox titrations and to determine the kappa number of wood pulp. It is mostly used in water treatment and as a disinfectant. It also shows useful application as a chemiluminescence reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only